Proceedings of the National Academy of Sciences of the United States of America
Induction of endonuclease-mediated apoptosis in tumor cells by C-nitroso-substituted ligands of poly(ADP-ribose) polymerase.
PMID 1502187
6-Nitroso-1,2-benzopyrone and 3-nitrosobenzamide, two C-nitroso compounds that inactivate the eukaryotic nuclear protein poly(ADP-ribose) polymerase [NAD+:poly(adenosine diphosphate D-ribose) ADP-D-ribosyltransferase, ADPRT, EC] at one zinc-finger site, completely suppressed the proliferation of leukemic and other malignant human cells and subsequently produced cell death. Tumoricidal concentrations of the drugs were relatively harmless to normal bone marrow progenitor cells and to superoxide formation by neutrophil granulocytes. The cellular mechanism elicited by the C-nitroso compounds consists of apoptosis due to DNA degradation by the nuclear calcium/magnesium-dependent endonuclease. This endonuclease is maintained in a latent form by poly(ADP-ribosyl)ation, but inactivation of ADPRT by C-nitroso drugs derepresses the DNA-degrading activity. ADPRT is thus identified as a critical regulatory enzyme component of a DNA-binding multiprotein system that plays a central function in defining DNA structures in the intact cell.