EMAIL THIS PAGE TO A FRIEND
Oncogene
SOCS1 [corrected] inhibits HPV-E7-mediated transformation by inducing degradation of E7 protein.
PMID 15021916
Abstract
Human papilloma viruses (HPVs) are small double-stranded DNA viruses that infect mucosal and cutaneous epithelium and induce cervical cancer. It has been shown that interferon (IFN)gamma suppresses proliferation of HPV-infected cells by suppressing expression of HPV E7. Here, we found that IFNgamma induces not only suppression of E7 transcription but also proteasome-dependent degradation. Suppressor of cytokine signaling-1 (SOCS1)/JAB, a suppressor of cytokine signaling, is known to be induced by IFNgamma, and functions as an antioncogene against various hematopoietic oncogenic proteins. SOCS1 contains the SOCS-box, which is shown to recruit ubiquitin transferase to the molecules that interact with SOCS1. We found that SOCS1 interacted with HPV E7 protein and induced ubiquitination and degradation of E7 in a SOCS-box-dependent manner. SOCS1 overexpression also increased Rb protein levels and suppressed proliferation of cervical cancer cell lines infected with HPV. Moreover, E7 protein levels were higher and Rb protein levels were lower in SOCS1-deficient fibroblasts infected with retrovirus vector carrying E7 gene than in wild-type fibroblasts. E7 induced anchorage-independent growth in SOCS1-deficient fibroblasts, but not in wild-type cells. These data suggested that SOCS1 plays an important role in regulating the levels of E7 protein and their transforming potential, and could be a new therapeutic tool for HPV-mediated tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04-002
Anti-SOCS1 Antibody, clone 4H1, clone 4H1, Upstate®, from mouse