EMAIL THIS PAGE TO A FRIEND
Human mutation
Biochemical characterization of two GALK1 mutations in patients with galactokinase deficiency.
PMID 15024738
Abstract
Galactokinase (GALK1) deficiency is an autosomal recessive disorder, which causes cataract formation in children not maintained on a lactose-free diet. Galactokinase deficiency results from mutation in the GALK1 gene mapped on 17q24. Since GK1 cDNA was cloned about 20 mutations (prevalently deletions and missense) have been reported to date. Most of these reported mutations are confined to single families, and only one of them, P28T, has been referred as the founder Romani mutation. In this paper we report two novel missense mutations in GALK1 gene, identified in two unrelated patients with galactokinase deficiency. One mutation, g.575G>A, substitutes a valine for a methionine at amino acid 32 (p.V32M), while the other mutation, g.2839G>A, results in the arginine to glutamine substitution p.R239Q (GenBank sequence L76927). Biochemical studies demonstrate that these mutations led to a drastic modification in GALK activity when individual mutant cDNAs were expressed in an E. coli system. These findings indicate the pathogeneticity of these mutations causing GALK deficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6048 Galactokinase human, recombinant, expressed in E. coli