EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
4'-pivaloyl substituted thymidine as a precursor for the thymyl radical: an EPR spectroscopic study.
PMID 15038294
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T72605
Trimethylacetyl chloride, 99%
C5H9ClO