EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
An improved total synthesis of triciribine: a tricyclic nucleoside with antineoplastic and antiviral properties.
PMID 15043134
Abstract
We describe an efficient total synthesis of triciribine, a tricyclic nucleoside with antineoplastic and antiviral properties, starting from 4-amino-6-bromo-5-cyanopyrrolo[2,3-d]pyrimidine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3580
Toyocamycin, ≥98% (HPLC), from Streptomyces rimosus
C12H13N5O4
T3830
Triciribine hydrate, ≥97% (HPLC)
C13H16N6O4 · xH2O