EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Synthesis of nucleoside libraries on solid support. II. 2,6,8-Trisubstituted purine nucleosides using 8-bromoguanosine as precursor.
PMID 15043170
Abstract
A series of 2,6,8-trisubstituted purine nucleoside libraries was prepared by parallel solid-phase synthesis using 8-bromoguanosine as a common synthetic precursor. Polystyrene-methoxytrityl chloride resin was linked to the N2 or O5' position of the guanosine analogues. 8-Bromoguanosine was derivatized at the C8 position via carbon-carbon bond formation. Nucleophilic aromatic substitution at C2 and/or C6 positions with various amines produced two series of purine nucleoside libraries with very diverse substitution.