EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Patterning of DNA nanostructures on silicon surface by electron beam lithography of self-assembled monolayer.
PMID 15045063
Abstract
Nanoscale patterns of modified oligonucleotides are produced on octadecyltrimethoxysilane self-assembled monolayers at a silicon surface by electron beam lithography. DNA structures with feature sizes of the order of 250 nm were detected by epi-fluorescence microscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
376213
Trimethoxy(octadecyl)silane, technical grade, 90%
C21H46O3Si