EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Coenzyme Q cytoprotective mechanisms.
PMID 15047098
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7956 Coenzyme Q1, ≥95%
C14H18O4