EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Aza-peptide Michael acceptors: a new class of inhibitors specific for caspases and other clan CD cysteine proteases.
PMID 15055989
Abstract
Aza-peptide Michael acceptors are a new class of irreversible inhibitors that are highly potent and specific for clan CD cysteine proteases. The aza-Asp derivatives were specific for caspases, while aza-Asn derivatives were effective legumain inhibitors. Aza-Lys and aza-Orn derivatives were potent inhibitors of gingipain K and clostripain. Aza-peptide Michael acceptors showed no cross reactivity toward papain, cathepsin B, and calpain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0888 Clostripain from Clostridium histolyticum, ≥20 units/mg solid