EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
The pharmacokinetics of tiopronin and its principal metabolite (2-mercaptopropionic acid) after oral administration to healthy volunteers.
PMID 1505618
Abstract
We have studied the pharmacokinetics of tiopronin and its principal metabolite, 2-mercaptopropionic acid (2-MPA) in healthy volunteers after the oral administration of 500 mg (2 Acadione tablets), followed by simultaneous assay of the two compounds in plasma over a period of 48 h using a new method (emission of fluorescence after HPLC and post-column derivatization by pyrene-maleimide). The absorption of tiopronin was slow (tmax between 4 and 6 h) and the plasma concentrations subsequently fell biexponentially. The principal metabolite 2-MPA appeared later in the plasma (tmax between 10 and 12 h after a lag-time of 3 h) then disappeared monoexponentially. About 15% of the tiopronin was metabolized to 2-MPA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W318000
2-Mercaptopropionic acid, ≥95%, FG
C3H6O2S
T31003
Thiolactic acid, 95%
C3H6O2S