EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Sensitive SPE-HPLC method to determine a novel angiotensin-AT1 antagonist in biological samples.
PMID 15063465
Abstract
A high-performance liquid chromatography (HPLC)-method after solid-phase extraction (SPE) has been developed in order to determine a new angiotensin-AT1 antagonist, i.e. CR 3210 (C27H24N8; MW = 460.54), 4-[4-[(2-ethyl-5,7-dimethylimidazo[4,5-b]pyridin-3-yl)methyl]phenyl]-3-(2H-tetrazol-5-yl)quinoline in rat plasma and urine after oral administration to Sprague-Dawley rats. CR 3210 and the internal standard (IS) CR 1505 (loxiglumide), i.e. 4-[(3,4-dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid, were isolated from rat urine and plasma by solid-phase extraction. The procedure was optimized regarding the sorbent extraction material, the pH in the conditioning solution, the washing step, the dry time and the type of elution solvent. The separation was performed by reversed-phase high-performance liquid chromatography with ultraviolet detection. The samples were injected onto the analytical column (Tracer Extrasil ODS1) and detected at 238 nm, giving a capacity factor of 1.87 for CR 3210 and 1.10 for the internal standard. The selectivity of the method was satisfactory. The mean recovery of CR 3210 from spiked rat plasma was 68.5 at 75 ng/ml and 80.9 at 3000 ng/ml; the mean recovery of CR 3210 from spiked rat urine was 69.9 at 75 ng/ml and 78.6 at 3000 ng/ml. The lower limit of detection (LOD) was 14 ng/ml in plasma and 22 ng/ml in urine samples. The lower limit of quantification (LOQ) was taken as 30 ng/ml, the lowest calibration standard using 500 microl rat plasma and urine. The procedures were validated according to international standards with a good reproducibility and linear response from 30 to 3000 ng/ml, for either plasma or urine. The sensitivity of the method allowed for its application to pharmacokinetic studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0130
Loxiglumide, ≥97% (HPLC)
C21H30Cl2N2O5