EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Cyclodextrin effect on solvolysis of substituted benzoyl chlorides.
PMID 15064797
Abstract
A kinetic study was carried out on the solvolysis of substituted benzoyl chlorides in the presence of alpha-, beta- and gamma-CD. Combination of the substituent dependent mechanism for solvolysis of benzoyl chlorides and the complexation ability of the cyclodextrin yields the following experimental behavior: (i) catalysis by beta- and gamma-CD for solvolysis of electron-attracting substituted benzoyl chlorides due to the reaction with its hydroxyl group C(6); (ii) absence of alpha-CD influence on solvolysis of benzoyl chlorides with electron withdrawing substituents; (iii) inhibition of solvolysis of benzoyl chlorides with electron-donating groups. This behavior is observed for solvolysis of meta/para substituted substrates in the presence of beta-CD, solvolysis of meta-substituted benzoyl chlorides in the presence of alpha-CD and solvolysis of para-substituted benzoyl chlorides in the presence of gamma-CD. This decrease in the rate constant is a consequence of the complexation of the substrate in the cyclodextrin cavity and its low solvation ability, causing the rate of solvolysis in its interior to be negligible. (iv) The solvolysis of meta-substituted benzoyl chlorides in the presence of gamma-CD yields a new behavior where the reaction of the complexed substrate is not negligible in the interior of the cyclodextrin cavity, which has been interpreted as a consequence of incomplete expulsion of hydration water from its cavity when the complexation takes place. (v) The experimental results obtained in the presence of alpha-CD show that meta-substituted benzoyl chlorides give rise to host : guest complexes with 1 : 1 stoichiometries, whereas those which are para-substituted cause a 2 : 1 stoichiometry to be formed. This difference in behavior has been interpreted taking into account the size of the different benzoyl chlorides and their accommodation in the alpha-CD cavity.