EMAIL THIS PAGE TO A FRIEND
Biochemistry
Tyrosine phosphorylation of the Janus kinase 2 activation loop is essential for a high-activity catalytic state but dispensable for a basal catalytic state.
PMID 15065871