EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Sodium dimercaptopropane sulfonate as antidote against non-metallic pesticides.
PMID 15066226
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31099
Chlordimeform, PESTANAL®, analytical standard
C10H13ClN2