EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Mammalian GLD-2 homologs are poly(A) polymerases.
PMID 15070731
Abstract
GLD-2 is a cytoplasmic poly(A) polymerase present in the Caenorhabditis elegans germ line and embryo. It is a divergent member of the DNA polymerase beta nucleotidyl transferase superfamily, which includes CCA-adding enzymes, DNA polymerases and eukaryotic nuclear poly(A) polymerases. The polyadenylation activity of GLD-2 is stimulated by physical interaction with an RNA binding protein, GLD-3. To test whether GLD-3 might stimulate GLD-2 by recruiting it to RNA, we tethered C. elegans GLD-2 to mRNAs in Xenopus oocytes by using MS2 coat protein. Tethered GLD-2 adds poly(A) and stimulates translation of the mRNA, demonstrating that recruitment is sufficient to stimulate polyadenylation activity. We use the same tethered assay to identify human and mouse poly(A) polymerases related to GLD-2. This may provide entrees to previously uncharacterized modes of polyadenylation in mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49115
Anti-PAPD4 antibody produced in rabbit, affinity isolated antibody