EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Reactions of methylenecyclopropanes with phenylsulfenyl chloride and phenylselenyl chloride.
PMID 15074931
Abstract
The reaction of methylenecyclopropanes 1 with phenylsulfenyl chloride or phenylselenyl chloride gives (cyclobut-1-enylsulfanyl)benzene or (cyclobut-1-enylselanyl)benzene along with ring-opened product in good total yields at 0 degrees C in various solvents. A plausible mechanism has been proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183342
Phenylselenyl chloride, 98%
C6H5ClSe