EMAIL THIS PAGE TO A FRIEND
Movement disorders : official journal of the Movement Disorder Society
Progressive supranuclear palsy syndrome induced by clebopride.
PMID 15077251
Abstract
We report on a patient who presented with a progressive supranuclear palsy (PSP) syndrome while receiving clebopride (CLB), a prokinetic drug with central antidopaminergic properties. The clinical and neurophysiological signs progressively disappeared after CLB withdrawal. To our knowledge, this is the first published PSP-like syndrome attributable to an antidopaminergic drug.