EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
TOX provides a link between calcineurin activation and CD8 lineage commitment.
PMID 15078895
Abstract
T cell development is dependent on the integration of multiple signaling pathways, although few links between signaling cascades and downstream nuclear factors that play a role in thymocyte differentiation have been identified. We show here that expression of the HMG box protein TOX is sufficient to induce changes in coreceptor gene expression associated with beta-selection, including CD8 gene demethylation. TOX expression is also sufficient to initiate positive selection to the CD8 lineage in the absence of MHC-TCR interactions. TOX-mediated positive selection is associated with up-regulation of Runx3, implicating CD4 silencing in the process. Interestingly, a strong T cell receptor-mediated signal can modify this cell fate. We further demonstrate that up-regulation of TOX in double positive thymocytes is calcineurin dependent, linking this critical signaling pathway to nuclear changes during positive selection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3501013
Anti-TOX antibody produced in rabbit, affinity isolated antibody