EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
A neutral organometallic fluoro complex can be a good ligand.
PMID 15079829
Abstract
The reaction of the complex [Mo(OTf)(eta(3)-C(3)H(4)-Me-2)(CO)(2)(phen)] (1) (OTf = trifluoromethylsulfonate; phen = 1,10-phenanthroline) with tetrabutylammonium fluoride trihydrate afforded the fluoride complex [MoF(eta(3)-C(3)H(4)-Me-2)(CO)(2)(phen)] (2). The IR spectrum and the oxidation potential of 2 reflect the fact that its metal center is more electron-rich than that of the chloro analogue [MoCl(eta(3)-C(3)H(4)-Me-2)(CO)(2)(phen)]. Compound 2 reacted with 1 affording the homobinuclear complex [[Mo(eta(3)-C(3)H(4)-Me-2)(CO)(2)(phen)](2)(mu-F)][OTf] (3), with a fluoride bridge. Compound 2 also reacts with the species generated in situ by triflate abstraction from [M(OTf)(CO)(3)('N-N')] (M = Mn, Re; 'N-N' = 2,2'-bipyridine (bipy), phen) using NaBAr'(4) (Ar' = 3,5-bis(trifluoromethyl)phenyl), affording the heterobinuclear complexes [[Mo(eta(3)-C(3)H(4)-Me-2)(CO)(2)(phen)](mu-F)[M(CO)(3)('N--N')]][BAr'(4)] (M = Mn, 'N-N' = bipy (4); M = Re, 'N-N' = phen (5)). All new compounds have been characterized by spectroscopic methods (IR and NMR) and, in the case of 1, 2, 3, and 4, also by means of X-ray diffraction analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86843
Tetrabutylammonium fluoride trihydrate, technical, ≥90% (T)
C16H36FN · 3H2O