EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Studies on the metabolism and the toxicological analysis of the nootropic drug fipexide in rat urine using gas chromatography-mass spectrometry.
PMID 15081928
Abstract
Qualitative studies are described on the metabolism and the toxicological analysis of the nootropic fipexide (FIP) in rat urine using gas chromatography-mass spectrometry (GC-MS). FIP was extensively metabolized to 1-(3,4-methylenedioxybenzyl)piperazine (MDBP), 4-chlorophenoxyacetic acid, 1-[2-(4-chlorophenoxy)acetyl]piperazine, N-(4-hydroxy-3-methoxy-benzyl)piperazine, piperazine, N-(3,4-methylenedioxybenzyl)ethylenediamine, and N-[2-(4-chlorophenoxy)acetyl]ethylenediamine. The authors' systematic toxicological analysis (STA) procedure using full-scan GC-MS after acid hydrolysis of one urine aliquot, liquid-liquid extraction and acetylation allowed the detection of FIP via its metabolites in rat urine after administration of a common FIP dose. Therefore, this qualitative procedure should also be suitable for detection of a FIP intake in human urine. Differentiation of an intake of FIP from that of other drugs which form common metabolites is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6270
Fipexide hydrochloride, analytical standard
C20H21ClN2O4 · HCl