EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
The effect of intravesical capsaicin and resiniferatoxin in neurogenic bladder dysfunction.
PMID 15088918
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R8756
Resiniferatoxin, from Euphorbia poisonii, ~95%
C37H40O9