EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Uptake of a fluorescent dye as a swift and simple indicator of organelle intactness: import-competent chloroplasts from soil-grown Arabidopsis.
PMID 15100247
Abstract
We developed a rapid and reliable technique for specifically staining intact chloroplasts using the fluorescent dye carboxyfluorescein diacetate. Intact, import-competent chloroplasts were isolated simply and rapidly from soil-grown Arabidopsis thaliana plants, with yields of 20 +/- 5 micro g chlorophyll per g FW, greater than previously reported yields from soil-grown Arabidopsis. Traditional chloroplast isolation buffers sometimes contain low concentrations (<10 mM) sodium ascorbate as a general-purpose anti-oxidant, but we found that only Arabidopsis chloroplasts isolated in the presence of high concentrations (50-100 mM) of sodium ascorbate in the initial grinding buffer were import-competent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4916
5-Carboxyfluorescein diacetate, ~95% (HPLC)
C25H16O9