EMAIL THIS PAGE TO A FRIEND
Acta anaesthesiologica Belgica
Substance use and misuse in pregnancy: peripartum anesthetic management of a parturient with recent methanol toluene and isopropanol intake.
PMID 15101148
Abstract
To date, no reports have specifically addressed the obstetric and anesthetic management of complications stemming from an acute maternal poly-inhalant intake; we herein present such a case and review the current limited literature available on this subject.