EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Microbial oxidation of adamantanone by Pseudomonas putida carrying the camphor catabolic plasmid.
PMID 1510672
Abstract
Intact cells of (+/-)camphor-grown Pseudomonas putida, ATCC17453(CAM), have been shown to oxidize readily the monoketone derivative of cage hydrocarbon adamantane, forming oxygenated products indicative of both biological Baeyer-Villiger and hydroxylation reactions. Formed products were identified as 4-oxahomoadamantan-5-one, 5-hydroxyadamantan-2-one and 1-hydroxy-4-oxahomoadamantan-5-one. Minor products formed as a result of secondary reactions were tentatively identified as syn- and anti-1,4-dihydroxyadamantanes and bicyclo[3.3.1]nonan-3-ol. Adamantanone initial concentrations determined whether 1-hydroxy-4-oxahomoadamantan-5-one was the sole product (below 120 mg/l) or 4-oxahomoadamantan-5-one was the principle (up to 92%) product (240-600 mg/l). Formation of 1-hydroxy-4-oxahomoadamantan-5-one appears to occur by two routes determined by the sequence of lactonization and hydroxylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
146048
2-Adamantanone, ReagentPlus®, 99%
C10H14O