EMAIL THIS PAGE TO A FRIEND
Emergency medicine journal : EMJ
Best evidence topic reports. Metoclopramide versus placebo with opioid.
PMID 15107376
Abstract
A short cut review was carried out to establish whether metoclopramide reduced nausea and vomiting after the administration of morphine. Altogether 405 papers were found using the reported search, of which one presented the best evidence to answer the clinical question. The author, date and country of publication, patient group studied, study type, relevant outcomes, results and study weaknesses of this best paper are tabulated. A clinical bottom line is stated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1824990
Metoclopramide, European Pharmacopoeia (EP) Reference Standard
C14H22ClN3O2