EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Efficient synthesis and structure-activity relationship of honokiol, a neurotrophic biphenyl-type neolignan.
PMID 15109665
Abstract
Honokiol, a biphenyl-type neolignan, which shows the remarkable neurotrophic effect in primary cultured rat cortical neurons, has been effectively synthesized in 21% yield over 14 steps starting from 5-bromosalicylic acid and p-hydroxybenzoic acid by utilizing Pd-catalyzed Suzuki-Miyaura coupling reaction as a key step. Additionally, the structure-activity relationship between neurite outgrowth-promoting activity and its O-methylated and/or its hydrogenated analogues was examined in the primary cultures of fetal rat cortical neurons, suggesting that 5-allyl and 4'-hydroxyl groups are essential for affecting the neurotrophic activity of honokiol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
461814
5-Bromosalicylic acid, technical grade, 90%
C7H5BrO3