EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Profound insights into squalene cyclization.
PMID 15112988
Abstract
In this issue of Chemistry & Biology, our understanding of the formation of pentacyclic hopene from the linear squalene is enhanced by an X-ray structure of a complex between squalene-hopene cyclase and the substrate analog 2-azasqualene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04626
Hop-22(29)-ene solution, 0.1 mg/mL in isooctane, analytical standard
C30H50