EMAIL THIS PAGE TO A FRIEND
Dermatologic therapy
Management of dyschromias in ethnic skin.
PMID 15113282
Abstract
Pigmentary disorders are one of the most common skin disorders among people of color. Dyspigmentation in the form of either hyperpigmentation or hypopigmentation is often psychologically devastating to patients with darker skin. There is marked contrast between normally pigmented hyperpigmented, hypopigmented or depigmented skin in people of color. Despite being common, pigmentary disorders remain difficult to treat.
Related Materials