EMAIL THIS PAGE TO A FRIEND
Pest management science
Insecticidal activity of chemical constituents from Aristolochia pubescens against Anticarsia gemmatalis larvae.
PMID 15119605
Abstract
Acetone and ethanol extracts of the tubercula and several compounds isolated from Aristolochia pubescens (Willd) were bioassayed on velvetbean caterpillars, Anticarsia gemmatalis (Hübner), for evaluation of the insecticidal activities. Of the extracts subjected to bioassay, the acetone extract showed the highest activity. (-)-Cubebin did not show activity against soybean caterpillars, whereas aristolochic acid and ent-kaur-15-en-17-ol increased the larval period. These compounds, and (+)-eudesmin and (+)-sesamin, reduced the viability of this period, giving rise to malformed adults. These extracts and compounds are therefore potential botanical insecticide agents for the control of velvetbean caterpillars in soybean crops.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00182
Eudesmine, ≥95% (LC/MS-ELSD)
C22H26O6