EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Crystallographic evidence for isomeric chromophores in 3-fluorotyrosyl-green fluorescent protein.
PMID 15122645
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F4505
m-Fluoro-DL-tyrosine
C9H10FNO3