EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Transforming chiral liquid chromatography methodologies into more sensitive liquid chromatography-electrospray ionization mass spectrometry without losing enantioselectivity.
PMID 15124813
Abstract
LC-electrospray ionization (ESI) MS conditions were optimized for the individual chiral separation of 19 compounds of pharmaceutical interest using the macrocyclic glycopeptide-based chiral stationary phases in both polar organic and reversed-phase modes (RPM). The influence of mobile phase composition and MS additive type on sensitivity was investigated for all classes of compounds tested. Compounds with amine or amide groups were efficiently separated, ionized, and detected with the addition of 0.1% (w/w) ammonium trifluoroacetate to the solvent system in either the reversed-phase or polar organic mode (POM). Macrocyclic glycopeptide coupled column technology was initially used to screen all chiral compounds analyzed. Baseline resolution of enantiomers was then achieved with relatively short retention times and high efficiencies on Chirobiotic T, Chirobiotic V or Chirobiotic R narrow bore chiral stationary phases. The polar organic mode offered better limits of detection (as low as 100 pg/ml) and sensitivity over reversed-phase methods. An optimum flow-rate range of 200-400 microl/min was necessary for sensitive chiral LC-ESI-MS analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13019AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
13020AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
13010AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
13022AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
13023AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
13024AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
12018AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
12019AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
12020AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
12010AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
12021AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
12022AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
12023AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
12024AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
12034AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
11018AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
11019AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
11020AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
11010AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
11022AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
11023AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
11024AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
11034AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
11044AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm