EMAIL THIS PAGE TO A FRIEND
Prikladnaia biokhimiia i mikrobiologiia
[Phenyl pyrazolones--novel oxidoreductase redox-mediators for degradation of xenobiotics].
PMID 15125193
Abstract
An approach was developed to screening organic compounds for putative activity of redox mediators of oxidoreductases, including laccases and peroxidases, applicable for xenobiotic degradation. The study was carried out with a homogenous laccase preparation from the basidiomycete Trametes hirsuta and horse-radish root peroxidase. Compounds belonging to 1-phenyl-3-methylpyrazolones were selected. Spectroscopic and electrochemical investigation of two of the compounds, sodium 1-phenyl-2,3-dimethyl-4-aminopyrazolon 5n(4)-methanesulfonate (PPNa) and 1-(3'-sulfophenyl)-3-methylpyrazolone (SPP), was performed. Electrochemical oxidation of both PPNa and SPP gave rise to high-potential intermediates capable of oxidizing veratryl alcohol; a lignin-modeling compound. Kinetic indices of these compounds were determined in enzymatic reactions with the presence of laccase. It was shown that enzymatic oxidation of SPP by laccase produced high-potential intermediates capable of oxidizing veratryl alcohol to veratric acid. Veratryl alcohol did not oxidize during enzymatic oxidation of SPP by peroxidase. This points to a difference between the mechanisms of enzymatic oxidation of PPNa and SPP by laccase and peroxidase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D131806
3,4-Dimethoxybenzoic acid, ≥99%
C9H10O4