EMAIL THIS PAGE TO A FRIEND
Prikladnaia biokhimiia i mikrobiologiia
[Properties and prospects of practical use of extracellular L-glutamate oxidase from Streptomyces sp. Z-11-6].
PMID 15125194
Abstract
The properties of extracellular L-glutamate oxidase, isolated and purified from Streptomyces sp. Z-11-6 (specific activity, 50.8 U/mg protein; yield, 40%), were studied. A photometrical method of determination of activities of alanine- and aspartate aminotransferases, based on the use of the L-glutamate oxidase and peroxidase, has been developed. This method is sufficiently sensitive to be used for the determination of aminotransferase activities in biological fluids. The presence of other amino acids did not interfere with the analysis and had no effect on the results of determination.