EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Tetrahydrogestrinone is a potent androgen and progestin.
PMID 15126583
Abstract
Tetrahydrogestrinone (THG) was recently identified as a novel steroid used illicitly to improve athletic performance. Although its structure is closely related to gestrinone, a 19-nor progestin, and resembles that of trenbolone, THG was never marketed, so information on its hormonal properties is not known. In this study, we demonstrate that THG is a highly potent androgen and progestin in a yeast-based in vitro bioassay system expressing human androgen and progesterone receptors. It has no estrogenic activity and no antagonism for any of the three steroid receptor classes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2321
Gestrinone, ≥97% (HPLC)
C21H24O2