EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly enantioselective construction of fused pyrrolidine systems that contain a quaternary stereocenter: concise formal synthesis of (+)-conessine.
PMID 15127448
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0095
Conessine, ≥97% (HPLC)
C24H40N2