EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Species and tissue differences in the toxicity of 3-butene-1,2-diol in male Sprague-Dawley rats and B6C3F1 mice.
PMID 15129020
Abstract
3-Butene-1,2-diol (BDD) is a major metabolite of 1,3-butadiene (BD), but the role of BDD in BD toxicity and carcinogenicity remains unclear. In this study, the acute toxicity of BDD was investigated in male Sprague-Dawley rats and B6C3F1 mice. Of the rats given 250 mg/kg BDD, 2 out of 4 died within 24 h; rats experienced hypoglycemia, significant alterations of liver integrity tests, and had lesions in the liver 4 h after treatment, but no lesions were detected in extrahepatic tissues. Rat hepatic GSH and GSSG levels were significantly depleted at both 1 and 4 h after the BDD treatment. Rats administered 200 mg/kg BDD also had liver lesions but no death or hypoglycemia was observed four or 24 h after treatment; these rats had depleted hepatic GSH and GSSG levels at 1 h but not at 4 or 24 h after treatment. Mice administered 250 mg/kg BDD exhibited modest alterations of liver integrity tests, but no death, hypoglycemia, or lesions in any tissue, and hepatic GSH and GSSG levels were depleted at 1 h but not at 4 h. The plasma half-life of BDD was four times longer in rats than in mice. Additional studies in rats showed the depletion of hepatic GSH and GSSG preceded the BDD-induced hypoglycemia and hepatotoxicity. Thus, the long half-life of BDD in rat plasma and the sustained depletion of hepatic GSH and GSSG may in part explain the higher sensitivity of the rat to BDD-induced hepatotoxicity. Furthermore, the results indicate that BDD may play a role in BD-induced toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
488216
3,4-Dihydroxy-1-butene, ≥99%
C4H8O2