EMAIL THIS PAGE TO A FRIEND
Epilepsy research
The effect of CGX-1007 and CI-1041, novel NMDA receptor antagonists, on NMDA receptor-mediated EPSCs.
PMID 15135163
Abstract
The N-methyl-D-aspartate (NMDA) receptor-gated ion channel is comprised of at least one NR1 subunit and any of four NR2 subunits (NR2A-D). The NR2 subunit confers different pharmacological and kinetic properties to the receptor. CGX-1007 (Conantokin G), a 17-amino acid polypeptide isolated from the venom of Conus geographus, is a novel NMDA receptor antagonist that is thought to be selective for the NR2B subunit. CGX-1007 has been reported to have highly potent, broad-spectrum anticonvulsant activity in animal seizure models. CI-1041 is an investigational compound, which also possesses anticonvulsant activity and has been shown to be highly selective for NR2B containing NMDA receptors. Although both CI-1041 and CGX-1007 are reportedly NR2B specific antagonists, they differ in their ability to block amygdala-kindled seizures, suggesting that the mechanism of action of these compounds differs. The present study was designed to test the hypothesis that CI-1041 and CGX-1007 would differentially modulate the function of NMDA receptors at excitatory synapses. Using the whole cell patch clamp technique, CGX-1007 and CI-1041 were found to block CA1 pyramidal cell, NMDA receptor-mediated excitatory postsynaptic currents (N-EPSCs) in a concentration-dependent manner in hippocampal slices from P4-P6 animals. In contrast, only CGX-1007 decreased NMDA receptor-mediated EPSC peak amplitude in slices from adult animals. The CGX-1007 block of peak amplitude was accompanied by a similar concentration-dependent decrease in decay kinetics of NMDA receptor-mediated EPSCs. These results suggest that while CI-1041 may be selective for NMDA receptors containing the NR2B subunit, CGX-1007 appears to be less selective than previously reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4311
Conantokin G, ≥75% (HPLC)
C88H139N26O44