EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
An Au-catalyzed cyclialkylation of electron-rich arenes with epoxides to prepare 3-chromanols.
PMID 15137751
Abstract
A gold-catalyzed cyclialkylation of electron-rich arenes with tethered epoxides afforded 3-chromanols stereospecifically.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
137995
1,2,3-Trimethoxybenzene, 98%
C9H12O3