EMAIL THIS PAGE TO A FRIEND
Cell Motility and the Cytoskeleton
C2C12 co-culture on a fibroblast substratum enables sustained survival of contractile, highly differentiated myotubes with peripheral nuclei and adult fast myosin expression.
PMID 15146538