EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
N1-(2-hydroxyethylthioethyl)-4-methyl imidazole (4-met-1-imid-thiodiglycol) in plasma and urine: a novel metabolite following dermal exposure to sulphur mustard.
PMID 1514930
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBR00018
2-(2-Methyl-1H-imidazol-1-yl)ethanol, AldrichCPR
C6H10N2O