EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Cryptobia iubilans infection in juvenile discus.
PMID 15154736
Abstract
Four commercial producers of discus (Symphysodon aequifasciatus) were found to have fish infested with the flagellate Cryptobia iubilans. Affected fish had granulomatous gastritis, and many also had granulomatous disease of other organs. The parasite had to be differentiated from the related flagellates Spironucleus spp, which induce different lesions. Transmission electron microscopy was found to be useful in detecting and identifying the parasite. Morbidity and mortality rates in the various fish populations appeared to be linked to a number of variables, including water quality, presence of other parasites and bacteria, diet, species, size, and age of the fish, and optimization of husbandry appeared to be important in alleviating the severity of disease. Metronidazole was not effective for treatment of C iubilans, but bath treatments with dimetridazole (80 mg/L for 24 hours, repeated daily for 3 days) or 2-amino-5-nitrothiazol (10 mg/L for 24 hours, repeated daily for 3 days) may be useful in decreasing the prevalence of infestation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4025
Dimetridazole
C5H7N3O2
31707
Dimetridazole, VETRANAL, analytical standard
C5H7N3O2