EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
Characteristics of the trigeminal depressor response in cats
PMID 15160400
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1585
Anti-Dopamine β Hydroxylase Antibody, serum, clone, Chemicon®