EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of 6'-S-alpha-D-glucopyranosyl-6'-thiomaltose (6(2)-thiopanose) and its effect on the enzymic activity of glucoamylases from Aspergillus niger.
PMID 1516093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2407
D-Panose, ≥97%
C18H32O16