EMAIL THIS PAGE TO A FRIEND
The Hematology Journal : The Official Journal of the European Haematology Association / EHA
Chronic lymphocytic leukaemia: new biological markers for assessing prognosis.
PMID 15167904