EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Synthesis and antibacterial activity of 7-(7-aminomethyl-5-azaspiro [2,4] hept-5-yl)-1-cyclopropyl-6-fluoro-8-methoxy-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid and its analogues].
PMID 15171652
Abstract
To find new antibacterial agents of quinolone with high activity and low toxicity. To design and synthesize 7-(7-aminomethyl-5-azaspiro [2,4] hept-5-yl)-1-cyclopropyl-6-fluoro-8-methoxy-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid and its analogues, and to study their antibacterial activity in vitro and in vivo. Twenty new compounds (2 - 11, 17 - 26) were obtained including five targeted compounds (22 - 26). The structures of the compounds were confirmed by 1HNMR, MS and HRMS. Compounds 22 - 26 showed broad spectrum of antibacterial activity against Gram-positive and Gram-negative organisms. Especially for compound 24, the relevant MIC values for 13 strains of Gram-positive organisms were < 0.001 - 0.03 mg(-1), including 4 strains of S. pneumoniae, 2 strains of S. pyogenes, 3 strains of S. aureus and 2 strains of Enterococci which exhibited more potent activity than contrast agents (clinafloxacin and gatifloxacin). The MIC values of 24 for 6 strains Gram-positive organisms were 0.01 - 1 mg x L(-1), which exhibited equal or lower activity than contrast agents. They were more effective than ciprofloxacin and gatifloxacin against intraperitoneal infections caused by S. pneumoniae and S. aureus in mice. Compounds (23, 24 and 26) showed excellent antibacterial activity in vitro and in vivo and should be worth further investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32144
Clinafloxacin, VETRANAL, analytical standard
C17H17ClFN3O3