EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Trafficking of Lyn through the Golgi caveolin involves the charged residues on alphaE and alphaI helices in the kinase domain.
PMID 15173188
Abstract
Src-family kinases, known to participate in signaling pathways of a variety of surface receptors, are localized to the cytoplasmic side of the plasma membrane through lipid modification. We show here that Lyn, a member of the Src-family kinases, is biosynthetically transported to the plasma membrane via the Golgi pool of caveolin along the secretory pathway. The trafficking of Lyn from the Golgi apparatus to the plasma membrane is inhibited by deletion of the kinase domain or Csk-induced "closed conformation" but not by kinase inactivation. Four residues (Asp346 and Glu353 on alphaE helix, and Asp498 and Asp499 on alphaI helix) present in the C-lobe of the kinase domain, which can be exposed to the molecular surface through an "open conformation," are identified as being involved in export of Lyn from the Golgi apparatus toward the plasma membrane but not targeting to the Golgi apparatus. Thus, the kinase domain of Lyn plays a role in Lyn trafficking besides catalysis of substrate phosphorylation.