EMAIL THIS PAGE TO A FRIEND
Science's STKE : Signal Transduction Knowledge Environment
From cell death to neuronal regeneration, effects of the p75 neurotrophin receptor depend on interactions with partner subunits.
PMID 15173600