EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Biosynthesis of volatiles by the myxobacterium Myxococcus xanthus.
PMID 15174160
Abstract
The volatiles emitted from cell cultures of myxobacterium Myxococcus xanthus were collected by use of a closed-loop stripping apparatus (CLSA) and analyzed by GC-MS. Two new natural products, (S)-9-methyldecan-3-ol ((S)-1) and 9-methyldecan-3-one (2), were identified and synthesized, together with other aliphatic ketones and alcohols, and terpenes. Biosynthesis of the two main components (S)-1 and 2 was examined in feeding experiments carried out with the wild-type strain DK1622 and two mutant strains JD300 and DK11017, which are impaired in the degradation pathway from leucine to isovaleryl-SCoA. Isovaleryl-SCoA is used as a starter, followed by chain elongation with two malonate units. Subsequent use of methyl malonate and decarboxylation leads to (S)-1 and 2. Furthermore, 3,3-dimethylacrylic acid (DMAA) can be used by the mutant strain to form isovaleryl-SCoA, which corroborates recent data on the detection of a novel variety of the mevalonate pathway giving rise to isovaleryl-SCoA from HMGCoA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136441
Dimethyl malonate, 98%
C5H8O4
63380
Dimethyl malonate, purum, ≥96.0% (GC)
C5H8O4