EMAIL THIS PAGE TO A FRIEND
Genes & development
Repression of the Arf tumor suppressor by E2F3 is required for normal cell cycle kinetics.
PMID 15175242
Abstract
Tumor development is dependent upon the inactivation of two key tumor-suppressor networks, p16(Ink4a)-cycD/cdk4-pRB-E2F and p19(Arf)-mdm2-p53, that regulate cellular proliferation and the tumor surveillance response. These networks are known to intersect with one another, but the mechanisms are poorly understood. Here, we show that E2F directly participates in the transcriptional control of Arf in both normal and transformed cells. This occurs in a manner that is significantly different from the regulation of classic E2F-responsive targets. In wild-type mouse embryonic fibroblasts (MEFs), the Arf promoter is occupied by E2F3 and not other E2F family members. In quiescent cells, this role is largely fulfilled by E2F3b, an E2F3 isoform whose function was previously undetermined. E2f3 loss is sufficient to derepress Arf, triggering activation of p53 and expression of p21(Cip1). Thus, E2F3 is a key repressor of the p19(Arf)-p53 pathway in normal cells. Consistent with this notion, Arf mutation suppresses the activation of p53 and p21(Cip1) in E2f3-deficient MEFs. Arf loss also rescues the known cell cycle re-entry defect of E2f3(-/-) cells, and this correlates with restoration of appropriate activation of classic E2F-responsive genes. Our data also demonstrate a direct role for E2F in the oncogenic activation of Arf. Specifically, we observe recruitment of the endogenous activating E2Fs, E2F1, and E2F3a, to the Arf promoter. Thus, distinct E2F complexes directly contribute to the normal repression and oncogenic activation of Arf. We propose that monitoring of E2F levels and/or activity is a key component of Arf's ability to respond to inappropriate, but not normal cellular proliferation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-603
Anti-Luciferase Antibody, clone21, clone 21, Upstate®, from mouse