EMAIL THIS PAGE TO A FRIEND
FEBS letters
The large subunit determines catalytic specificity of barley sucrose:fructan 6-fructosyltransferase and fescue sucrose:sucrose 1-fructosyltransferase.
PMID 15178325
Abstract
Plant fructosyltransferases are highly homologous in primary sequence and typically consist of two subunits but catalyze widely different reactions. Using functional expression in the yeast Pichia pastoris, we show that the substrate specificity of festuca sucrose:sucrose 1--beta-D-fructosyltransferase (1-SST) and barley sucrose:fructan 6--beta-D-fructosyltransferase (6-SFT) is entirely determined by the large subunit. Chimeric enzymes with the large subunit of festuca 1-SST (LSuB) and the small subunit of barley 6-SFT have the same catalytic specificity as the native festuca 1-SST and vice versa. If the LSuB is expressed alone, it does not yield a functionally active enzyme, indicating that the small subunit is nevertheless essential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56218
Nystose, analytical standard
C24H42O21